Anti-CCNL1 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8459-55EA 4320 DKK
USBIC8459-55
Anti-CCNL1 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:5000-1:25000; Western Blot: 1:2000-1:5000; Immunochemistry: 10-20 ?g/ml

Type: Primary
Antigen: CCNL1 (cyclin L1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR