Anti-CCNDBP1 Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8456-01BEA 4120 DKK
USBIC8456-01B
Anti-CCNDBP1 Goat polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:32000; Immunochemistry: 3-6?g/ml

Type: Primary
Antigen: CCNDBP1 (cyclin D-type binding-protein 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Dog, Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR