Anti-Cyclin E2, Isoform 3 Chicken Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8455-36EA 4290 DKK
USBIC8455-36
Anti-Cyclin E2, Isoform 3 Chicken Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: Cyclin E2, Isoform 3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR