Anti-CCNE1 Rabbit monoclonal antibody [clone: 9H224]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8455-26HEA 4000 DKK
USBIC8455-26H
Anti-CCNE1 Rabbit monoclonal antibody [clone: 9H224]
Antistoffer
Recommended dilution: Flow Cytometry: 1:20; Western Blot: 1:1000-1:2000; Immunochemistry: 1:100-1:250; Immunoprecipitation: 1:80

Type: Primary
Antigen: CCNE1 (cyclin E1)
Clonality: Monoclonal
Clone: 9H224
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR