Anti-CCNE1 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8455-26BEA 2590 DKK
USBIC8455-26B
Anti-CCNE1 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Immunochemistry:1:50-100

Type: Primary
Antigen: CCNE1 (cyclin E1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR