Anti-CCNE1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBIC8455-01K-APCEA 6360 DKK
USBIC8455-01K-APC
Anti-CCNE1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antistoffer
Recommended dilution: Immunochemistry: 1:50-1:100; Flow Cytrometry: 1:10-1:50; Western Blot: 1:50-1:100
Do not freeze!
Type: Primary
Antigen: CCNE1 (cyclin E1)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human