Anti-CCNE1 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8455-01HEA 4190 DKK
USBIC8455-01H
Anti-CCNE1 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:2000; Western Blot: 1:200-1:500

Type: Primary
Antigen: CCNE1 (cyclin E1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR