Anti-DMTF1 Mouse monoclonal antibody HRP (Horseradish Peroxidase) [clone: 10C10]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8454-80B-HRPEA 0 DKK
USBIC8454-80B-HRP
Anti-DMTF1 Mouse monoclonal antibody HRP (Horseradish Peroxidase) [clone: 10C10]
Type: Primary
Antigen: DMTF1 (cyclin D binding myb-like transcription factor 1)
Clonality: Monoclonal
Clone: 10C10
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR