Anti-DMTF1 Mouse Monoclonal Antibody [clone: 8C92]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8454-80EA 2530 DKK
USBIC8454-80
Anti-DMTF1 Mouse Monoclonal Antibody [clone: 8C92]
Antistoffer
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: DMTF1 (cyclin D binding myb-like transcription factor 1)
Clonality: Monoclonal
Clone: 8C92
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR