Anti-CCND3 Mouse monoclonal antibody [clone: 11n57]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8454-25BEA 4320 DKK
USBIC8454-25B
Anti-CCND3 Mouse monoclonal antibody [clone: 11n57]
Antistoffer
Recommended dilution: Western Blot: 1 ?g/ml; Immunochemistry: 8-25 ?g/ml

Type: Primary
Antigen: CCND3 (cyclin D3)
Clonality: Monoclonal
Clone: 11n57
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR