Anti-CCND3 Mouse monoclonal antibody [clone: 10K179]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8454-20CEA 3290 DKK
USBIC8454-20C
Anti-CCND3 Mouse monoclonal antibody [clone: 10K179]
Antistoffer
Recommended dilution: Western Blot: 0.25-0.5 ?g/ml; Immunochemistry: 0.5-1 ?g/ml

Type: Primary
Antigen: CCND3 (cyclin D3)
Clonality: Monoclonal
Clone: 10K179
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR