Anti-CCND2 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8454-12JEA 3590 DKK
USBIC8454-12J
Anti-CCND2 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 1 ?g/ml

Type: Primary
Antigen: CCND2 (cyclin D2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR