Anti-CCND2 Mouse monoclonal antibody [clone: 0.N.240]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8454-12EEA 2850 DKK
USBIC8454-12E
Anti-CCND2 Mouse monoclonal antibody [clone: 0.N.240]
Antistoffer
Type: Primary
Antigen: CCND2 (cyclin D2)
Clonality: Monoclonal
Clone: 0.N.240
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR