Anti-CCND1 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8454-03BEA 2620 DKK
USBIC8454-03B
Anti-CCND1 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 1 ?g/ml; Immunohistology: 1:100

Type: Primary
Antigen: CCND1 (cyclin D1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR