Anti-CCND1 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8454-02LEA 4030 DKK
USBIC8454-02L
Anti-CCND1 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 4-6?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: CCND1 (cyclin D1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR