Anti-CCND1 Mouse monoclonal antibody [clone: 10K177]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8454-02EEA 3290 DKK
USBIC8454-02E
Anti-CCND1 Mouse monoclonal antibody [clone: 10K177]
Antistoffer
Recommended dilution: Western Blot: 4?g/ml; Immunochemistry: 8?g/ml

Type: Primary
Antigen: CCND1 (cyclin D1)
Clonality: Monoclonal
Clone: 10K177
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR