Anti-CCND1 Mouse monoclonal antibody [clone: 7H91]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8454-02DEA 4320 DKK
USBIC8454-02D
Anti-CCND1 Mouse monoclonal antibody [clone: 7H91]
Antistoffer
Recommended dilution: Immunochemistry: 10 ?g/ml; ELISA: 0.5-1 ?g/ml

Type: Primary
Antigen: CCND1 (cyclin D1)
Clonality: Monoclonal
Clone: 7H91
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR