Anti-CCND1 Rabbit polyclonal antibody APC (Allophycocyanin)

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8454-01S-APCEA 0 DKK
USBIC8454-01S-APC
Anti-CCND1 Rabbit polyclonal antibody APC (Allophycocyanin)
Type: Primary
Antigen: CCND1 (cyclin D1)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR