Anti-CCND1 Mouse monoclonal antibody [clone: CD1.1]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8454-01B2EA 4320 DKK
USBIC8454-01B2
Anti-CCND1 Mouse monoclonal antibody [clone: CD1.1]
Antistoffer
Recommended dilution: Western Blot: 1 ?g/ml; Immunochemistry: 1-10 ?g/ml;

Type: Primary
Antigen: CCND1 (cyclin D1)
Clonality: Monoclonal
Clone: CD1.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR