Anti-CCNC Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8453-32EA 3650 DKK
USBIC8453-32
Anti-CCNC Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 1:100-1:200; Immunoprecipitation: 10?l/mg

Type: Primary
Antigen: CCNC (cyclin C)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR