Anti-CCNB2 Mouse monoclonal antibody [clone: 8C91]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8453-28EA 2530 DKK
USBIC8453-28
Anti-CCNB2 Mouse monoclonal antibody [clone: 8C91]
Antistoffer
Type: Primary
Antigen: CCNB2 (cyclin B2)
Clonality: Monoclonal
Clone: 8C91
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR