Anti-CCNB1 Rabbit monoclonal antibody [clone: 8A11]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8453-04DEA 4550 DKK
USBIC8453-04D
Anti-CCNB1 Rabbit monoclonal antibody [clone: 8A11]
Antistoffer
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:100

Type: Primary
Antigen: CCNB1 (cyclin B1)
Clonality: Monoclonal
Clone: 8A11
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR