Anti-CCNB1 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8453-02EA 3350 DKK
USBIC8453-02
Anti-CCNB1 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 1:1000; Immunofluorescence: 1:25

Type: Primary
Antigen: CCNB1 (cyclin B1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Hamster, Human, Monkey, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR