Anti-CCNB1 Rabbit Polyclonal Antibody (Biotin)

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8453-01J-BIOTEA 6360 DKK
USBIC8453-01J-BIOT
Anti-CCNB1 Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: CCNB1 (cyclin B1)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR