Anti-CCNA2 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8452-72GEA 3810 DKK
USBIC8452-72G
Anti-CCNA2 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 5 ?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: CCNA2 (cyclin A2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR