Anti-CCNA1 Mouse polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8452-70EA 2860 DKK
USBIC8452-70
Anti-CCNA1 Mouse polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 1:500-1:1000

Type: Primary
Antigen: CCNA1 (cyclin A1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR