Anti-cGMP Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8450-25EEA 3310 DKK
USBIC8450-25E
Anti-cGMP Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: EIA: 1:10-1:50

Type: Primary
Antigen: cGMP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR