Anti-Cyclic Adenosine Monophosphate Sheep Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8450-12AEA 5540 DKK
USBIC8450-12A
Anti-Cyclic Adenosine Monophosphate Sheep Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 10.26?g/ml

Type: Primary
Antigen: Cyclic Adenosine Monophosphate
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR