Anti-ACKR3 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8376-26BEA 4320 DKK
USBIC8376-26B
Anti-ACKR3 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Flow Cytometry: 1-10 ?g/ml; Western Blot: 4-6?g/ml; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: ACKR3 (atypical chemokine receptor 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR