Anti-ACKR3 Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: 11n55]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8376-26AEA 5060 DKK
USBIC8376-26A
Anti-ACKR3 Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: 11n55]
Antistoffer
Type: Primary
Antigen: ACKR3 (atypical chemokine receptor 3)
Clonality: Monoclonal
Clone: 11n55
Conjugation: Peridinin Chlorophyll
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR