Anti-ACKR3 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 8B9]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8376-26EA 4590 DKK
USBIC8376-26
Anti-ACKR3 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 8B9]
Antistoffer
Recommended dilution: Immunochemistry: 8-25 ?g/ml

Type: Primary
Antigen: ACKR3 (atypical chemokine receptor 3)
Clonality: Monoclonal
Clone: 8B9
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR