Anti-ACKR3 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8376-25GEA 4320 DKK
USBIC8376-25G
Anti-ACKR3 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:500-1:1000; Immunofluorescence: 1:100-1:500; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: ACKR3 (atypical chemokine receptor 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR