Anti-ACKR3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7H134]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8376-25BEA 4500 DKK
USBIC8376-25B
Anti-ACKR3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7H134]
Antistoffer
Do not freeze!

Type: Primary
Antigen: ACKR3 (atypical chemokine receptor 3)
Clonality: Monoclonal
Clone: 7H134
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR