Anti-ACKR3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 11C11]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8376-25AEA 4970 DKK
USBIC8376-25A
Anti-ACKR3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 11C11]
Antistoffer
Do not freeze!

Type: Primary
Antigen: ACKR3 (atypical chemokine receptor 3)
Clonality: Monoclonal
Clone: 11C11
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR