Anti-CXCR6 Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: 11n52]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8375-13BEA 4590 DKK
USBIC8375-13B
Anti-CXCR6 Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: 11n52]
Antistoffer
Type: Primary
Antigen: CXCR6 (chemokine (C-X-C motif) receptor 6)
Clonality: Monoclonal
Clone: 11n52
Conjugation: Peridinin Chlorophyll
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR