Anti-CXCR4b Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8375-03EA 2750 DKK
USBIC8375-03
Anti-CXCR4b Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:8000

Type: Primary
Antigen: CXCR4b
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Zebrafish
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR