Anti-CXCR4 Rat Monoclonal Antibody (Peridinin Chlorophyll) [clone: 11C10]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8375-01JEA 4440 DKK
USBIC8375-01J
Anti-CXCR4 Rat Monoclonal Antibody (Peridinin Chlorophyll) [clone: 11C10]
Antistoffer
Do not freeze!

Type: Primary
Antigen: CXCR4 (chemokine (C-X-C motif) receptor 4)
Clonality: Monoclonal
Clone: 11C10
Conjugation: Peridinin Chlorophyll
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR