Anti-CXCR4 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8370-01EA 3210 DKK
USBIC8370-01
Anti-CXCR4 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 0.5-4 ?g/ml

Type: Primary
Antigen: CXCR4 (chemokine (C-X-C motif) receptor 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR