Anti-CXCR4 Mouse Monoclonal Antibody [clone: 5J286]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8352-01EA 4820 DKK
USBIC8352-01
Anti-CXCR4 Mouse Monoclonal Antibody [clone: 5J286]
Antistoffer
Type: Primary
Antigen: CXCR4 (chemokine (C-X-C motif) receptor 4)
Clonality: Monoclonal
Clone: 5J286
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR