Anti-CXCR4 Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8350-08DEA 4340 DKK
USBIC8350-08D
Anti-CXCR4 Goat Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:200-300

Type: Primary
Antigen: CXCR4 (chemokine (C-X-C motif) receptor 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR