Anti-CXCR4 Mouse Monoclonal Antibody [clone: 7H160]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8350-06CEA 4320 DKK
USBIC8350-06C
Anti-CXCR4 Mouse Monoclonal Antibody [clone: 7H160]
Antistoffer
Recommended dilution: Flow Cytometry: 25 ?g/ml; Immunochemistry: 25 ?g/ml

Type: Primary
Antigen: CXCR4 (chemokine (C-X-C motif) receptor 4)
Clonality: Monoclonal
Clone: 7H160
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Cat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR