Anti-CXCR4 Mouse Monoclonal Antibody [clone: 12G5]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBIC8350-05CEA 4730 DKK
USBIC8350-05C
Anti-CXCR4 Mouse Monoclonal Antibody [clone: 12G5]
Antistoffer
Recommended dilution: Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: CXCR4 (chemokine (C-X-C motif) receptor 4)
Clonality: Monoclonal
Clone: 12G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human