Anti-CXCR4-Lo Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8350-02AEA 3340 DKK
USBIC8350-02A
Anti-CXCR4-Lo Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 5-1 ?g/ml

Type: Primary
Antigen: CXCR4-Lo
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR