Anti-CCR4 Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8350-01QEA 4320 DKK
USBIC8350-01Q
Anti-CCR4 Goat polyclonal antibody
Antistoffer
Recommended dilution: Flow Cytometry: 1:12ELISA: 1:30000; Western Blot: 1:500; Immunochemistry: 1:200

Type: Primary
Antigen: CCR4 (chemokine (C-C motif) receptor 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR