Anti-CCR4 Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: 11D80]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8350-01GEA 4780 DKK
USBIC8350-01G
Anti-CCR4 Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: 11D80]
Antistoffer
Do not freeze!

Type: Primary
Antigen: CCR4 (chemokine (C-C motif) receptor 4)
Clonality: Monoclonal
Clone: 11D80
Conjugation: Peridinin Chlorophyll
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR