Anti-CXCR-3.2 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8349-07EA 2900 DKK
USBIC8349-07
Anti-CXCR-3.2 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:5000-1:20000

Type: Primary
Antigen: CXCR-3.2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Zebrafish
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR