Anti-CXCR2 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8348-04CEA 2300 DKK
USBIC8348-04C
Anti-CXCR2 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 0. 05-0.2?g/ml; Western Blot: 0.5-2?g/ml; Immunochemistry: 5-10 ?g/ml

Type: Primary
Antigen: CXCR2 (chemokine (C-X-C motif) receptor 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR