Anti-CX3CR1, EL Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8300-01AEA 4840 DKK
USBIC8300-01A
Anti-CX3CR1, EL Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1:500; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: CX3CR1, EL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR