Anti-CX3CR1 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8299-01AEA 2720 DKK
USBIC8299-01A
Anti-CX3CR1 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1:1000-1:2000; Flow Cytometry: 1:100-1:300; Immunochemistry: 1:100-1:300

Type: Primary
Antigen: CX3CR1 (chemokine (C-X3-C motif) receptor 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR