Anti-CXCL16 Goat Polyclonal Antibody (Biotin)

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC8298-04B-50EA 2900 DKK
USBIC8298-04B-50
Anti-CXCL16 Goat Polyclonal Antibody (Biotin)
Antistoffer
Recommended dilution: ELISA: 0.15-0. 30 ?g/ml

Type: Primary
Antigen: CXCL16 (chemokine (C-X-C motif) ligand 16)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR